Stampa…

N-(4-Cyanophenyl)guanidine ≥99.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-Cyanophenyl)guanidine

C3387-1G C3387-5G
TCIAC3387-1GEA 26 EUR
TCIAC3387-1G TCIAC3387-5G
N-(4-Cyanophenyl)guanidine ≥99.0% (by HPLC, total nitrogen)
N-(4-Cyanophenyl)guanidine
Formula: C₈H₈N₄
Peso molecolare: 160,18 g/mol
MDL Number: MFCD16251323
CAS Number: 5637-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd31cb4a23453e3c38031d129e0728dfa614817f&vid=cd31cb4a23453e3c38031d129e0728dfa614817f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point (Decomposition) 210.0 to 214.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR