1-Cyclopentenylboronic acid pinacol ester ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-(Cyclopent-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopentene, 1-Cyclopenteneboronic acid Pinacol ester, 2-(1-Cyclopentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C3385-1G
C3385-5G
TCIAC3385-1GEA
33
EUR
TCIAC3385-1G
TCIAC3385-5G
1-Cyclopentenylboronic acid pinacol ester ≥98.0% (by GC)
1-Cyclopentenylboronic acid pinacol ester
Formula:
C₁₁H₁₉BO₂ Peso molecolare: 194,0784 g/mol |
MDL Number:
MFCD08669391 CAS Number: 287944-10-9 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...