Stampa…

4-Chloro-2-(trifluoromethoxy)aniline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylamine

C3374-1G C3374-200MG
TCIAC3374-1GEA 73 EUR
TCIAC3374-1G TCIAC3374-200MG
4-Chloro-2-(trifluoromethoxy)aniline ≥98.0% (by GC)
4-Chloro-2-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
Peso molecolare: 211,57 g/mol
MDL Number: MFCD00153102
CAS Number: 175205-77-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR