Stampa…

2-Chloro-4-iodo-3-methylpyridine ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 2-Chloro-4-iodo-3-picoline

C3368-1G C3368-5G
TCIAC3368-1GEA 52 EUR
TCIAC3368-1G TCIAC3368-5G
2-Chloro-4-iodo-3-methylpyridine ≥98.0% (by GC)
2-Chloro-4-iodo-3-methylpyridine
Formula: C₆H₅ClIN
MDL Number: MFCD03095293
CAS Number: 153034-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a047986db5728634e73b9e79d49f0a25cbe80866&vid=a047986db5728634e73b9e79d49f0a25cbe80866&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR