Stampa…

5-Chloro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Chloropyridine-3-boronic acid Pinacol ester, 2-(5-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

C3367-200MG C3367-1G
TCIAC3367-200MGEA 20 EUR
TCIAC3367-200MG TCIAC3367-1G
5-Chloro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
5-Chloro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
Peso molecolare: 239,51 g/mol
MDL Number: MFCD06798284
CAS Number: 865186-94-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR