Stampa…

6-Chloro-2-iodo-3-hydroxypyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloro-2-iodopyridin-3-ol

C3366-1G C3366-5G
TCIAC3366-1GEA 69 EUR
TCIAC3366-1G TCIAC3366-5G
6-Chloro-2-iodo-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
6-Chloro-2-iodo-3-hydroxypyridine
Formula: C₅H₃ClIO
Peso molecolare: 255,44 g/mol
MDL Number: MFCD07636749
CAS Number: 188057-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68915be0f9dd913be593e1009be25b2fa71aeeed&vid=68915be0f9dd913be593e1009be25b2fa71aeeed&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR