Stampa…

3-Chloro-5-fluorobenzaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C3365-1G C3365-5G
TCIAC3365-1GEA 35 EUR
TCIAC3365-1G TCIAC3365-5G
3-Chloro-5-fluorobenzaldehyde ≥98.0% (by GC)
3-Chloro-5-fluorobenzaldehyde
Formula: C₇H₄ClFO
Peso molecolare: 158,56 g/mol
MDL Number: MFCD02261734
CAS Number: 90390-49-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR