Stampa…

[4'-(Carbazol-9-yl)-4-biphenylyl]boronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4'-(9H-Carbazol-9-yl)-[1,1'-biphenyl]-4-yl)boronic acid

C3353-200MG C3353-1G
TCIAC3353-200MGEA 50 EUR
TCIAC3353-200MG TCIAC3353-1G
[4'-(Carbazol-9-yl)-4-biphenylyl]boronic acid (contains varying amounts of Anhydride)
[4'-(Carbazol-9-yl)-4-biphenylyl]boronic acid
Formula: C₂₄H₁₈BNO₂
Peso molecolare: 363,22 g/mol
MDL Number: MFCD28978624
CAS Number: 858131-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3fffa5a59ccefd97d613c1537db0e4acd9c4767&vid=b3fffa5a59ccefd97d613c1537db0e4acd9c4767&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 95.0 to 105.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR