Stampa…

4-Chloro-3-fluorobenzylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (4-Chloro-3-fluorophenyl)methanamine

C3344-1G C3344-5G
TCIAC3344-1GEA 84 EUR
TCIAC3344-1G TCIAC3344-5G
4-Chloro-3-fluorobenzylamine ≥98.0% (by GC, titration analysis)
4-Chloro-3-fluorobenzylamine
Formula: C₇H₇ClFN
Peso molecolare: 159,59 g/mol
MDL Number: MFCD04116008
CAS Number: 72235-58-6
UN: 2735
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR