4-(Chloromethyl)-4'-methyl-2,2'-bipyridyl ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
4-(Chloromethyl)-4'-methyl-2,2'-bipyridine
Formula:
C₁₂H₁₁ClN₂ |
MDL Number:
MFCD24464190 CAS Number: 83799-54-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...