Stampa…

tert-Butyl carbamate ≥98.0% (by GC)

Fornitore: TCI

B3132-25G B3132-5G
TCIAB3132-25GEA 65 EUR
TCIAB3132-25G TCIAB3132-5G
tert-Butyl carbamate ≥98.0% (by GC)
tert-Butyl carbamate
Formula: C₅H₁₁NO₂
MDL Number: MFCD00007962
CAS Number: 4248-19-5
EINECS: 224-209-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a4fbc793c43055e1575c234c01ca7a5ae9c2d1b&vid=4a4fbc793c43055e1575c234c01ca7a5ae9c2d1b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR