Stampa…

2-(4-Chlorophenyl)-2-propanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Chloro-alpha,alpha-dimethylbenzyl alcohol

C3324-1G C3324-5G
TCIAC3324-1GEA 121 EUR
TCIAC3324-1G TCIAC3324-5G
2-(4-Chlorophenyl)-2-propanol ≥98.0% (by GC)
2-(4-Chlorophenyl)-2-propanol
Formula: C₉H₁₁ClO
Peso molecolare: 170,64 g/mol
Punto di ebollizione: 96°C
MDL Number: MFCD11870102
CAS Number: 1989-25-9
EINECS: 217-865-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR