Stampa…

6-Chloro-4-quinolinol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloroquinolin-4-ol, 6-Chloro-4-hydroxyquinoline

C3312-1G C3312-5G
TCIAC3312-1GEA 37 EUR
TCIAC3312-1G TCIAC3312-5G
6-Chloro-4-quinolinol ≥98.0% (by GC)
6-Chloro-4-quinolinol
Formula: C₉H₆ClNO
Peso molecolare: 179,60 g/mol
Punto di fusione: 269°C
MDL Number: MFCD00024011
CAS Number: 23432-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098bf7e195ec0ccef1bc38438e83ee6ca1703052&vid=098bf7e195ec0ccef1bc38438e83ee6ca1703052&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR