Stampa…

(3-Chloro-4-methoxyphenyl)acetic acid ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 3-Chloro-4-methoxyphenylacetic acid, 2-(3-Chloro-4-methoxyphenyl)acetic acid

C3311-1G C3311-5G
TCIAC3311-1GEA 99 EUR
TCIAC3311-1G TCIAC3311-5G
(3-Chloro-4-methoxyphenyl)acetic acid ≥97.0% (by GC)
(3-Chloro-4-methoxyphenyl)acetic acid
Formula: C₉H₉ClO₃
Peso molecolare: 200.62 g/mol
MDL Number: MFCD02664697
CAS Number: 13721-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11bfbb3d4b6847e6b77288e85da9bee812f90726&vid=11bfbb3d4b6847e6b77288e85da9bee812f90726&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 92.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR