(2-Carboxyethyl)triphenylphosphonium bromide ≥96.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₂₁H₂₀BrO₂P |
MDL Number:
MFCD00031698 CAS Number: 51114-94-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...