Stampa…

3-Chloro-5-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Chloro-5-cyanobenzotrifluoride

C3307-5G C3307-25G
TCIAC3307-5GEA 73 EUR
TCIAC3307-5G TCIAC3307-25G
3-Chloro-5-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
3-Chloro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
Peso molecolare: 205,57 g/mol
MDL Number: MFCD04115854
CAS Number: 693245-52-2
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d8360d242185c8f87b263730b5215bcef3387f&vid=66d8360d242185c8f87b263730b5215bcef3387f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR