4'-Chloro-3'-fluoroacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(4-Chloro-3-fluorophenyl)ethanone
C3298-1G
C3298-5G
TCIAC3298-1GEA
39
EUR
TCIAC3298-1G
TCIAC3298-5G
4'-Chloro-3'-fluoroacetophenone ≥98.0% (by GC)
4'-Chloro-3'-fluoroacetophenone
Formula:
C₈H₆ClFO Peso molecolare: 172,59 g/mol |
MDL Number:
MFCD04115858 CAS Number: 151945-84-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...