Stampa…

3-Carboxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Carboxy-5-fluorobenzeneboronic acid, 3-Borono-5-fluorobenzoic acid

C3287-1G C3287-5G
TCIAC3287-1GEA 58 EUR
TCIAC3287-1G TCIAC3287-5G
3-Carboxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Carboxy-5-fluorophenylboronic acid
Formula: C₇H₆BFO₄
Peso molecolare: 183,93 g/mol
MDL Number: MFCD07363791
CAS Number: 871329-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcd2c81fda0a20d2c4bca256ecd99f40d5017866&vid=dcd2c81fda0a20d2c4bca256ecd99f40d5017866&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 95.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR