Stampa…

(E)-4'-Chloro-4-methoxychalcone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (E)-4'-Chloro-2-(4-methoxybenzal)acetophenone, (E)-4'-Chloro-2-(4-methoxybenzylidene)acetophenone

C3280-1G C3280-5G
TCIAC3280-1GEA 99 EUR
TCIAC3280-1G TCIAC3280-5G
(E)-4'-Chloro-4-methoxychalcone ≥98.0% (by GC)
(E)-4'-Chloro-4-methoxychalcone
Formula: C₁₆H₁₃ClO₂
Peso molecolare: 272,73 g/mol
MDL Number: MFCD00018708
CAS Number: 85502-87-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR