4'-Cyano-4-decyloxybiphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
10OCB, 4'-Decyloxy-4-biphenylcarbonitrile, 4'-(Decyloxy)-[1,1'-biphenyl]-4-carbonitrile
C3269-1G
C3269-5G
TCIAC3269-1GEA
89
EUR
TCIAC3269-1G
TCIAC3269-5G
4'-Cyano-4-decyloxybiphenyl ≥98.0% (by GC)
4'-Cyano-4-decyloxybiphenyl
Formula:
C₂₃H₂₉NO Peso molecolare: 335,49 g/mol |
MDL Number:
MFCD01218034 CAS Number: 70247-25-5 EINECS: 274-495-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 79.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...