Cesium(I) bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Bis(trifluoromethanesulphonyl)imide Cesium(I) salt, Cesium(I) triflimide
Formula:
C₂CsF₆NO₄S₂ Punto di fusione: 122°C |
MDL Number:
MFCD29472520 CAS Number: 91742-16-4 UN: 1759 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...