2-Chloro-6-fluorophenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(2-Chloro-6-fluorophenyl)boronic acid, (2-Chloro-6-fluorophenyl)metaboric acid, 2-Chloro-6-fluorobenzeneboronic acid, Dihydroxy(2-chloro-6-fluorophenyl)borane
C3247-1G
C3247-5G
TCIAC3247-1GEA
30
EUR
TCIAC3247-1G
TCIAC3247-5G
2-Chloro-6-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-6-fluorophenylboronic acid
Formula:
C₆H₅BClFO₂ Peso molecolare: 174,37 g/mol |
MDL Number:
MFCD04039892 CAS Number: 313545-32-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...