4-Carboxy-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Borono-2-nitrobenzoic acid, 3-Nitro-4-carboxyphenylboronic acid, 4-Carboxy-3-nitrobenzeneboronic acid
C3233-200MG
C3233-1G
TCIAC3233-200MGEA
32
EUR
TCIAC3233-200MG
TCIAC3233-1G
4-Carboxy-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-3-nitrophenylboronic acid
Formula:
C₇H₆BNO₆ Peso molecolare: 210,94 g/mol Punto di fusione: 220°C |
MDL Number:
MFCD10696661 CAS Number: 80500-28-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 98.0 to 109.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...