Stampa…

Cyproheptadine hydrochloride sesquihydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Cyproheptadine hydrochloride, 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methylpiperidine hydrochloride

C3218-5G C3218-25G
TCIAC3218-5GEA 40 EUR
TCIAC3218-5G TCIAC3218-25G
Cyproheptadine hydrochloride sesquihydrate ≥98.0% (by HPLC, titration analysis)
Cyproheptadine hydrochloride sesquihydrate
Formula: C₂₁H₂₁N·HCl·11/2H₂O
Peso molecolare: 350,88 g/mol
MDL Number: MFCD00012538
CAS Number: 41354-29-4
EINECS: 213-535-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,02773

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (after drying)
Drying loss 7.0 to 9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR