Stampa…

2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Cyano-N-(4-(trifluoromethyl)phenyl)acetamide

C3217-1G C3217-5G
TCIAC3217-1GEA 107 EUR
TCIAC3217-1G TCIAC3217-5G
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide ≥98.0% (by GC)
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Formula: C₁₀H₇F₃N₂O
Peso molecolare: 228,17 g/mol
MDL Number: MFCD05669245
CAS Number: 24522-30-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR