Stampa…

2-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

C3213-200MG C3213-1G
TCIAC3213-200MGEA 64 EUR
TCIAC3213-200MG TCIAC3213-1G
2-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
Peso molecolare: 224,3726 g/mol
MDL Number: MFCD09258745
CAS Number: 957061-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45f1644941a6ac7a63760e81f0c316f0de530110&vid=45f1644941a6ac7a63760e81f0c316f0de530110&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR