Stampa…

5-Chloro-2-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Chloro-2-ethoxybenzeneboronic acid

C3210-1G C3210-5G
TCIAC3210-1GEA 24 EUR
TCIAC3210-1G TCIAC3210-5G
5-Chloro-2-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
5-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
Peso molecolare: 200,43 g/mol
MDL Number: MFCD03427055
CAS Number: 352534-86-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR