Stampa…
1-Chloro-2,4-bis(hexyloxy)benzene ≥96.0% (by GC)
Fornitore: TCI
C3206-1G C3206-5G
TCIAC3206-1GEA 53 EUR
TCIAC3206-1G TCIAC3206-5G
1-Chloro-2,4-bis(hexyloxy)benzene ≥96.0% (by GC)
1-Chloro-2,4-bis(hexyloxy)benzene
Formula: C₁₈H₂₉ClO₂
MDL Number: MFCD29089351
CAS Number: 851228-26-7
Visualizza i codici
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %