Cinnamylphosphonic acid ≥98.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₉H₁₁O₃P |
MDL Number:
MFCD19105641 CAS Number: 146404-58-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 170.0 to 175.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...