4'-Chloro-2,2':6',2''-terpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
C3187-200MG
C3187-1G
TCIAC3187-200MGEA
38
EUR
TCIAC3187-200MG
TCIAC3187-1G
4'-Chloro-2,2':6',2''-terpyridine ≥98.0% (by GC, titration analysis)
4'-Chloro-2,2':6',2''-terpyridine
Formula:
C₁₅H₁₀ClN₃ Peso molecolare: 267,716 g/mol |
MDL Number:
MFCD00191930 CAS Number: 128143-89-5 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...