Stampa…

3-Chloro-5-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C3183-1G C3183-5G
TCIAC3183-1GEA 47 EUR
TCIAC3183-1G TCIAC3183-5G
3-Chloro-5-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Chloro-5-hydroxybenzoic acid
Formula: C₇H₅ClO₃
MDL Number: MFCD04114327
CAS Number: 53984-36-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 244.0 to 248.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR