1-Chloro-2-naphthol ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
1-Chloronaphthalen-2-ol
Formula:
C₁₀H₇ClO Punto di ebollizione: 157°C |
MDL Number:
MFCD00997084 CAS Number: 633-99-8 EINECS: 211-200-4 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...