Stampa…

2-(4'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI
Attenzione

C3164-200MG C3164-1G
TCIAC3164-200MGEA 26 EUR
TCIAC3164-200MG TCIAC3164-1G
2-(4'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(4'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₇H₁₈ClN₃
MDL Number: MFCD28975108
CAS Number: 1443049-85-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 218.0 to 222.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR