Stampa…

1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1-(4-Chlorophenyl)-4,4,4-trifluorobutane-1,3-dione, 3-(4-Chlorobenzoyl)-1,1,1-trifluoroacetone

C3139-1G C3139-5G
TCIAC3139-1GEA 33 EUR
TCIAC3139-1G TCIAC3139-5G
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione ≥98.0% (by GC, titration analysis)
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
Formula: C₁₀H₆ClF₃O₂
Peso molecolare: 250,61 g/mol
MDL Number: MFCD00511276
CAS Number: 18931-60-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 60.0 to 64.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR