(3-Chloro-4-fluorophenyl)thiourea ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
C3137-1G
C3137-5G
TCIAC3137-1GEA
64
EUR
TCIAC3137-1G
TCIAC3137-5G
(3-Chloro-4-fluorophenyl)thiourea ≥98.0% (by HPLC)
(3-Chloro-4-fluorophenyl)thiourea
Formula:
C₇H₆ClFN₂S Peso molecolare: 204,65 g/mol |
MDL Number:
MFCD01571081 CAS Number: 154371-25-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...