6-Chloro-7-iodo-7-deazapurine ≥95.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine
C3130-1G
C3130-5G
TCIAC3130-1GEA
34
EUR
TCIAC3130-1G
TCIAC3130-5G
6-Chloro-7-iodo-7-deazapurine ≥95.0% (by GC, titration analysis)
6-Chloro-7-iodo-7-deazapurine
Formula:
C₆H₃ClIN₃ Peso molecolare: 279,47 g/mol |
MDL Number:
MFCD09263258 CAS Number: 123148-78-7 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 200.0 to 204.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...