Stampa…

2-Chloro-4,6-diphenylpyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C3123-1G C3123-5G
TCIAC3123-1GEA 52 EUR
TCIAC3123-1G TCIAC3123-5G
2-Chloro-4,6-diphenylpyrimidine ≥98.0% (by GC)
2-Chloro-4,6-diphenylpyrimidine
Formula: C₁₆H₁₁ClN₂
Peso molecolare: 266,73 g/mol
MDL Number: MFCD00030776
CAS Number: 2915-16-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR