5-Chloro-2-fluorobenzaldehyde ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
C3114-5G
C3114-25G
TCIAC3114-5GEA
91
EUR
TCIAC3114-5G
TCIAC3114-25G
5-Chloro-2-fluorobenzaldehyde ≥98.0% (by GC)
5-Chloro-2-fluorobenzaldehyde
Formula:
C₇H₄ClFO Peso molecolare: 158.56 g/mol |
MDL Number:
MFCD03094518 CAS Number: 96515-79-6 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 35.0 to 39.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...