1-(cyclopropylcarbonyl)piperazine hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cyclopropyl(piperazin-1-yl)methanone hydrochloride, 1-(Cyclopropylcarbonyl)piperazine hydrochloride
C3112-1G
C3112-5G
TCIAC3112-1GEA
53
EUR
TCIAC3112-1G
TCIAC3112-5G
1-(cyclopropylcarbonyl)piperazine hydrochloride ≥98.0% (by titrimetric analysis)
1-(cyclopropylcarbonyl)piperazine hydrochloride
Formula:
C₈H₁₅ClN₂O Peso molecolare: 190,671 g/mol |
MDL Number:
MFCD03428576 CAS Number: 1021298-67-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...