Stampa…

Cefdinir ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

C3111-1G C3111-5G
TCIAC3111-1GEA 92 EUR
TCIAC3111-1G TCIAC3111-5G
Cefdinir ≥97.0% (by HPLC, titration analysis)
Cefdinir
Formula: C₁₄H₁₃N₅O₅S₂
Punto di fusione: 190°C
MDL Number: MFCD00865030
CAS Number: 91832-40-5
Merck Index: 14,01920

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D -58.0 to -66.0 deg (C=1,0.1mol/L buffer sol. pH7.0 Phosphate)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR