2-Chloro-3-nitro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
C3089-1G
C3089-5G
TCIAC3089-1GEA
44
EUR
TCIAC3089-1G
TCIAC3089-5G
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Formula:
C₆H₂ClF₃N₂O₂ Peso molecolare: 226,54 g/mol |
MDL Number:
MFCD06255171 CAS Number: 72587-15-6 |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...