4-Chloro-2-(methylthio)pyrimidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Chloro-2-methylthiopyrimidine
C3087-5G
C3087-25G
TCIAC3087-5GEA
42
EUR
TCIAC3087-5G
TCIAC3087-25G
4-Chloro-2-(methylthio)pyrimidine ≥98.0% (by GC)
4-Chloro-2-(methylthio)pyrimidine
Formula:
C₅H₅ClN₂S Peso molecolare: 160,63 g/mol Punto di ebollizione: 112°C |
MDL Number:
MFCD00006083 CAS Number: 49844-90-8 EINECS: 256-500-6 UN: 1760 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...