Stampa…

3-(4-Chlorobutyl)indole-5-carbonitrile ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Chlorobutyl)-1H-indole-5-carbonitrile, 3-(4-Chlorobutyl)-5-cyanoindole

C3086-1G C3086-5G
TCIAC3086-1GEA 37 EUR
TCIAC3086-1G TCIAC3086-5G
3-(4-Chlorobutyl)indole-5-carbonitrile ≥97.0% (by GC)
3-(4-Chlorobutyl)indole-5-carbonitrile
Formula: C₁₃H₁₃ClN₂
Peso molecolare: 232,71 g/mol
MDL Number: MFCD09833261
CAS Number: 143612-79-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR