Stampa…

3-Chloro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C3083-1G C3083-5G
TCIAC3083-1GEA 28 EUR
TCIAC3083-1G TCIAC3083-5G
3-Chloro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
3-Chloro-4-hydroxybenzonitrile
Formula: C₇H₄ClNO
Peso molecolare: 153,57 g/mol
MDL Number: MFCD01567246
CAS Number: 2315-81-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR