3-Chloro-2-ethoxypyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
C3081-1G
C3081-5G
TCIAC3081-1GEA
93
EUR
TCIAC3081-1G
TCIAC3081-5G
3-Chloro-2-ethoxypyridine ≥98.0% (by GC, titration analysis)
3-Chloro-2-ethoxypyridine
Formula:
C₇H₈ClNO Peso molecolare: 157,60 g/mol |
MDL Number:
MFCD09702462 CAS Number: 177743-06-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...