2-Chloroisonicotinamide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
C3079-1G
C3079-5G
TCIAC3079-1GEA
29
EUR
TCIAC3079-1G
TCIAC3079-5G
2-Chloroisonicotinamide ≥98.0% (by titrimetric analysis)
2-Chloroisonicotinamide
Formula:
C₆H₅ClN₂O |
MDL Number:
MFCD00221401 CAS Number: 100859-84-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 196.0 to 200.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...