Stampa…

3'-Chloro-4'-hydroxyacetophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(3-Chloro-4-hydroxyphenyl)ethanone, 3-Chloro-4-hydroxyacetophenone, 1-(3-Chloro-4-hydroxyphenyl)ethan-1-one

C3076-1G C3076-5G
TCIAC3076-1GEA 37 EUR
TCIAC3076-1G TCIAC3076-5G
3'-Chloro-4'-hydroxyacetophenone ≥98.0% (by GC)
3'-Chloro-4'-hydroxyacetophenone
Formula: C₈H₇ClO₂
Peso molecolare: 170,6 g/mol
MDL Number: MFCD00663621
CAS Number: 2892-29-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR