Stampa…
6-Chloroflavone ≥98.0% (by GC)
Fornitore: TCI
Attenzione
C3066-1G C3066-5G
TCIAC3066-1GEA 74 EUR
TCIAC3066-1G TCIAC3066-5G
6-Chloroflavone ≥98.0% (by GC)
6-Chloroflavone
Formula: C₁₅H₉ClO₂
MDL Number: MFCD00209560
CAS Number: 10420-73-2
Visualizza i codici
Purity (GC) min. 98.0 %
Melting point 183.0 to 187.0 deg-C