Stampa…

Carbinoxamine Maleate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: Carbinoxamine maleate salt

C3057-5G C3057-25G
TCIAC3057-5GEA 38 EUR
TCIAC3057-5G TCIAC3057-25G
Carbinoxamine Maleate ≥98.0% (by HPLC, titration analysis)
Carbinoxamine Maleate
Formula: C₁₆H₁₉ClN₂O·C₄H₄O₄
Peso molecolare: 406.86 g/mol
MDL Number: MFCD00082461
CAS Number: 3505-38-2
EINECS: 222-498-0
UN: 2811
ADR: 6.1 III
Merck Index: 14,01799

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR