Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine ≥97.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
Synonyms:
5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Copper(II)
C3051-200MG
C3051-1G
TCIAC3051-200MGEA
118
EUR
TCIAC3051-200MG
TCIAC3051-1G
Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine ≥97.0% (by total nitrogen and titration analysis)
Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine
Formula:
C₈₀H₈₈CuN₈O₈ Peso molecolare: 1353,18 g/mol |
MDL Number:
MFCD00191950 CAS Number: 155773-67-4 |
Risultati del test di specificazione
Appearance | Red to Dark blue to Black powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...